Learn More
Medchemexpress LLC Butyrylcholinesteras 1mg
SDP
Supplier:  Medchemexpress LLC HYP728591MG
Encompass_Preferred
Butyrylcholinesterase (BCHE) Protein an esterase with broad substrate specificity crucially inactivates acetylcholine maintaining neurotransmitter homeostasis Beyond acetylcholine metabolism BCHE degrades neurotoxic organophosphates highlighting its detoxification role The enzyme s versatility underscores its critical function in safeguarding against harmful effects of neurotoxic compounds Butyrylcholinesterase/BCHE Protein Mouse (HEK293 His) is the recombinant mouse-derived Butyrylcholinesterase/BCHE protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-81825