Learn More
Medchemexpress LLC Butyryltimolol | 106351-79-5 | 98.0% | 386.51 g/mol | C17H30N4O4S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1020325MG
Encompass_Preferred
Butyryltimolol is a lipophilic ester prodrug of timolol that functions as a β-adrenergic blocker and is used in research to improve corneal penetration and ocular absorption of timolol. It is supplied as a high-purity research reagent with documented solubility and storage recommendations.
Catalog No. 50-002-64122