Learn More
Medchemexpress LLC Butyryltimolol | 106351-79-5 | MFCD32898870 | 98.0% | 386.51 g/mol | C17H30N4O4S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10203225MG
Encompass_Preferred
Butyryltimolol is a β-adrenergic blocker and lipophilic ester prodrug of timolol that improves corneal penetration. Supplied for research use, it is available as a solid and in solution formats with specified purity and storage recommendations; handle per supplier safety and storage guidance.
  • Acts as a β-adrenergic blocker and timolol precursor.
  • Improves corneal penetration compared with timolol.
  • Reported purity: 98.0%.
  • Molecular formula C17H30N4O4S; molecular weight 386.51 g/mol.
  • Available as solid packs and as a 10 mM solution in DMSO; typical pack sizes 5-500 mg.
  • For research use only; observe storage conditions for pure material and solutions.
Catalog No. 50-002-47668