Learn More
Medchemexpress LLC Buvacitam impurity 27 | 50840-31-8 | MFCD18785533 | 97.0% | 120.58 g/mol | C4H9ClN2 | 10 MG

Supplier: Medchemexpress LLC HYW01893610MG
Buvacitam impurity 27 (3-aminobutanenitrile hydrochloride, CAS 50840-31-8) is a characterized synthetic impurity supplied in a 10 mg laboratory pack for research and analytical use. It is intended as an impurity reference for method development, impurity profiling, and analytical validation.
- Contains 10 mg of material.
- Chemical formula C4H9ClN2; molecular weight 120.58 g/mol.
- Typical purity reported as 97.0%.
- Provided for research use only; not for human or therapeutic use.
- Safety data sheet and batch certificate of analysis available.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.