Learn More
ACCEGEN BIOTECH BV-2,q Frozen, 1 vial
Supplier:  ACCEGEN BIOTECH ABCTC212S
Encompass
Tissue: brain, microglial cells; Transformed by: recombinant retrovirus (v-raf/v-mic). The BV-2 cells express the nuclear v-myc and the cytoplasmic v-raf oncogene products as well as the env gp70 antigen at the surface level; The BV-2 cells have morphological, phenotypical and functional markers of macrophages.
Catalog No. NC1826152