Learn More
Biolegend Brilliant Violet 421™ anti-mouse CD279 (PD-1)
Supplier:  Biolegend 135218
Encompass_Preferred
Brilliant Violet 421™ anti-mouse CD279 (PD-1) [29F.1A12]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 uL
Catalog No. 50-403-051