Learn More
Biolegend Brilliant Violet 570™ anti-mouse CD11c
Supplier:  Biolegend 117331
Encompass_Preferred
Brilliant Violet 570™ anti-mouse CD11c [N418]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. 50-163-721