Learn More
Biolegend Brilliant Violet 650™ anti-human CX3CR1
Supplier:  Biolegend 341626
Encompass_Preferred
Brilliant Violet 650™ anti-human CX3CR1 [2A9-1]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-6472