Learn More
Biolegend Brilliant Violet 650™ anti-mouse TER-119/Erythroid Cells
Supplier:  Biolegend 116235
Encompass_Preferred
Brilliant Violet 650™ anti-mouse TER-119/Erythroid Cells [TER-119]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 125 uL
Catalog No. NC1739193