Learn More
Biolegend Brilliant Violet 711™ anti-human CX3CR1
Supplier:  Biolegend 341630
Encompass_Preferred
Brilliant Violet 711™ anti-human CX3CR1 [2A9-1]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-6475