Learn More
Medchemexpress LLC BVT.13 | 756813-87-3 | 98.4% | 404.2 | C18H11Cl2N3O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1148535MG
Encompass_Preferred
BVT.13 is an orally active, selective peroxisome proliferator-activated receptor gamma (PPARγ) agonist used as a research compound. It displays a maximal efficacy similar to rosiglitazone and has demonstrated antidiabetic activity in ob/ob mice, making it useful for studies of PPARγ signaling, metabolic regulation, and preclinical efficacy testing.
  • Selective PPARγ agonist activity.
  • Oral bioactivity demonstrated in animal models.
  • Demonstrated antidiabetic efficacy in ob/ob mice.
  • High purity: 98.4% (HPLC).
  • Suitable for in vitro and in vivo research applications.
  • Molecular weight 404.2; formula C18H11Cl2N3O4.
Catalog No. 50-002-64428