Learn More
Selleck Chemical LLC BVT-948 S9829-5MG
SDP
Supplier:  Selleck Chemical LLC S98295MG
Encompass_Preferred
BVT 948 is a irreversible noncompetitive and cell-permeable protein tyrosine phosphatase PTP inhibitor with IC50s of 0 9 M 1 7 M 0 09 M 1 5 M and 0 7 M for PTP1B TCPTP SHP-2 LAR and YopH respectively
Catalog No. 50-000-24726