Learn More
Medchemexpress LLC BW-A 78U | 101155-02-6 | 99.7% | 257.27 | C13H12FN5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10011810MM 1ML
Encompass_Preferred
BW-A 78U is a small-molecule phosphodiesterase 4 (PDE4) inhibitor with a reported IC50 of 3 μM. It is supplied as a 10 mM solution in DMSO for biochemical and cell-based research applications.
  • Inhibits PDE4 with reported IC50 of 3 μM.
  • Provided as a ready-to-use 10 mM solution in DMSO.
  • Packaged as a 1 mL vial for small-scale experiments.
  • High purity (99.7%).
  • Suitable for biochemical assays and cell-based studies.
  • Molecular formula C13H12FN5 and molecular weight 257.27 Da.
Catalog No. 50-002-15181