Learn More
Medchemexpress LLC Bx795 | 702675-74-9 | 99.84% | 591.47 | 200 MG
Supplier: Medchemexpress LLC 200MGHY10514
BX795 is a potent and selective inhibitor of PDK1, with an IC50 of 6 nM. It is also a potent and relatively specific inhibitor of TBK1 and IKKε, having IC50 values of 6 nM and 41 nM, respectively. BX795 works by blocking the phosphorylation of S6K1, Akt, PKCδ, and GSK3β, and exhibits lower selectivity over PKA, PKC, c-Kit, and GSK3β, among others. Additionally, BX795 modulates autophagy.
- Effectively blocks PDK1 activity in PC-3 cells.
- Inhibits the phosphorylation of S6K1, Akt, PKCδ, and GSK3β.
- Potently inhibits tumor cell growth on plastic.
- Shows higher growth inhibition in soft agar.
- Blocks TBK1- and IKKε-mediated activation of IRF3 and the production of IFN-β.
- Demonstrates an inhibitory effect on 2-MeSADP-induced or collagen-induced aggregation, ATP secretion, and thromboxane generation in platelet physiological responses.