Learn More
Medchemexpress LLC HY-N6022 5mg Medchemexpress, Byakangelicin CAS:482-25-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN60225MG
Encompass_Preferred
Medchemexpress, HY-N6022 5mg Byakangelicin CAS:482-25-7 Byakangelicin, one of the active compounds found in the roots of Angelica gigas, can serve as a modulator to improve brain accumulation of diverse active compounds (Umb, Cur, and Dox) and enhance therapeutic effects. Byakangelicin is likely to increase the expression of all PXR target genes (such as MDR1) and induce a wide range of drug-drug interactions. Byakangelicin can inhibit the effects of sex hormones, it may increase the catabolism of endogenous hormones. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2944