Learn More
Medchemexpress LLC (±)-Byakangelicol | 26091-79-2 | 99.9% | 316.31 g·mol⁻¹ | C17H16O6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN007410MG
Encompass_Preferred
Byakangelicol is a naturally occurring phenylpropanoid isolated from Angelica dahurica that inhibits interleukin-1β-induced prostaglandin E2 (PGE2) production by suppressing cyclooxygenase-2 (COX-2) expression and activity. It is supplied for research use only.
  • Natural phenylpropanoid isolated from Angelica dahurica.
  • Inhibits COX-2 expression and activity, reducing PGE2 release.
  • Suitable for inflammation and immunology research applications.
  • Offered in small milligram pack sizes for assay development.
  • Certificate of analysis and purity documentation available.
Catalog No. 50-002-97788