Learn More
Cytek Biosciences Inc cFluor® BYG710 Anti-Human IgD (IgD26) - 25 Tests
Supplier:  Cytek Biosciences Inc R720392
Encompass_Preferred
The 4A11 monoclonal antibody binds to the Fab fragment of human IgG. IgG is a membrane BCR that is produced when activated naïve B cells undergo class switch recombination of the Ig heavy chain constant region. This class-switching results in IgM and IgD being replaced by IgG as the activated B cell develops into memory and antibody secreting cells. On memory B cells, when IgG binds to its target antigen, it initiates signaling and activation that rapidly leads to proliferation and differentiation into antibody secreting cells. The antibody was conjugated to a fluorophore and purified by chromatography.
Catalog No. NC2888862