Learn More
Accuflow Systems Saint-Gobain Bytac Bench Protector, FEP, 12.5 in x 5 yd
Supplier:  Accuflow Systems D9860012
Encompass_Preferred
The Bytac bench protector is a self-adhesive surface lining composed of an inert FEP fluoropolymer film bonded to an aluminum foil backing with a pressure-sensitive acrylic adhesive. It provides a chemically resistant, non-stick surface that protects laboratory benches, fume hoods, walls, and spray booths from spills, stains, and contamination. The roll is cut to size for simple application and removal, and wipes clean with a damp sponge.
  • FEP fluoropolymer surface resists corrosive chemicals, paint, oil, and grease.
  • Aluminum foil backing provides structural support and reflectivity.
  • Pressure-sensitive acrylic adhesive allows easy application and removal.
  • Roll format, 12.5 in wide, 5 yd length; cut to size with scissors.
  • Suitable for bench tops, fume hoods, walls, splash screens, and spray booths.
Catalog No. NC2327771