Learn More
Glen Research Bz-A-LA-CE Phosphoramidite, 10-2000-10E, Locked Nucleic Acid (LNA), 2' O-methylene linkage, 885.96 g/mol, Anhydrous Acetonitrile, Freezer storage -10 to -30°C, Stability 2-3 days
Supplier:  Glen Research 10200010E
Encompass_Preferred
Bz-A-LA-CE Phosphoramidite, 10-2000-10E, Locked Nucleic Acid (LNA), 2' O-methylene linkage, 885.96 g/mol, Anhydrous Acetonitrile, Freezer storage -10 to -30°C, Stability 2-3 days
Catalog No. NC3346898