Learn More
Bachem Bz-Arg-pNA · HCl
SDP
Supplier:  Bachem 4000792.0250
Encompass_Preferred
Bz-Arg-pNA · HCl, 21653-40-7, C19H22N6O4 · HCl, Mr 434.88, 250mg. L-BAPA · HCl, L-BApNA · HCl. Substrate for trypsin. BApNA is also cleaved by papain, acrosin, and plasmin.
Catalog No. 50-260-384