Learn More
Cayman Chemical Bz-FVR-AMCtrIfluoroacetate 1mg
SDP
Supplier:  Cayman Chemical 248451 MG
Encompass_Preferred
A fluorogenic peptide substrate; AMC is released upon enzymatic cleavage and its fluorescence can be used to quantify enzyme activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-293-5703