Learn More
Medchemexpress LLC Bz-(Me)Tz-NHS | 1454558-58-7 | 95.0% | 412.40 g/mol | C19H20N6O5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15168610MG
Encompass_Preferred
Bz-(Me)Tz-NHS is a methyltetrazine-containing N-hydroxysuccinimide (NHS) ester used as a bioorthogonal linker for copper-free click chemistry. It undergoes rapid inverse electron-demand Diels-Alder (iEDDA) reactions with strained alkenes to enable fast, selective conjugation under mild, biocompatible conditions. The reagent is supplied as a purple solid with high purity and requires sealed, low-temperature storage to maintain stability.
  • Methyltetrazine-containing NHS ester enabling copper-free inverse electron-demand Diels-Alder conjugation.
  • Rapid reaction kinetics with strained alkenes for efficient labeling.
  • NHS ester functional group for covalent coupling to primary amines.
  • High purity suitable for bioconjugation workflows.
  • Purple solid appearance for easy visual identification.
  • Stable when stored sealed at 4°C; in solvent store at -80°C for long-term preservation.
Catalog No. 50-002-87299