Learn More
Medchemexpress LLC Bz-rA (N6-Benzoyladenosine) | 4546-55-8 | MFCD00056988 | 371.35 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW008388
Encompass_Preferred
Bz-rA (N6-Benzoyladenosine) is a nucleoside derivative featuring an N6 protecting group. It is primarily utilized as a synthetic intermediate in photocatalytic oxidative cyclization reactions and for the synthesis of oligoribonucleotides. Bz-rA effectively facilitates the construction of heterocyclic skeletons in photochemical conversions and safeguards the ribose moiety to control the cyclization pathway.
  • Nucleoside derivative with an N6 protecting group
  • Utilized as a synthetic intermediate in photocatalytic oxidative cyclization reactions
  • Used for the synthesis of oligoribonucleotides
  • Facilitates the construction of heterocyclic skeletons in photochemical conversions
  • Safeguards the ribose moiety to control the cyclization pathway
Catalog No. 50-004-48169