Learn More
Medchemexpress LLC Bz-rA phosphoramidite | 104992-55-4 | 988.19 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0061021G
Encompass_Preferred
Bz-rA Phosphoramidite is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis. It is a 2'-O-protected ribonucleoside phosphoramidite.
  • Can be used in the synthesis of 2'-O-guanidinopropyl-modified nucleoside phosphoramidites.
Catalog No. 50-004-46220