Learn More
Selleck Chemical LLC C-171-E0128-5MG
SDP
Supplier:  Selleck Chemical LLC E01285MG
Encompass_Preferred
C-171 is a covalent small-molecule inhibitor of STING C-171 efficiently inhibits both hsSTING and mmSTING through covalently target the predicted transmembrane cysteine residue 91 and thereby block the activation-induced palmitoylation of STING
Catalog No. 50-234-0853