Learn More
Medchemexpress LLC Octanamide, N-[(1S,2R,3E)-2-hydroxy-1-[(phosphonooxy)methyl]-3-heptadecen-1-yl]- | 158983-53-0 | 99.0% | 505.67 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1650995MG
Encompass_Preferred
C-8 Ceramide-1-phosphate is a chemical compound primarily identified for its role in cellular processes. It functions as an inhibitor of cell apoptosis and an inducer of cell survival.
  • Inhibits cell apoptosis
  • Induces cell survival
  • Available in an off-white to light yellow solid form
  • High purity of 99.0%
  • Stable storage conditions for powder and in solvent
Catalog No. 50-003-91444