Learn More
ABclonal Technology c-Abl Rabbit mAb
SDP
Supplier:  ABclonal Technology A2208220UL
Encompass_Preferred
This gene is a protooncogene that encodes a protein tyrosine kinase involved in a variety of cellular processes including cell division adhesion differentiation and response to stress The activity of the protein is negatively regulated by its SH3 domain whereby deletion of the region encoding this domain results in an oncogene The ubiquitously expressed protein has DNA-binding activity that is regulated by CDC2-mediated phosphorylation suggesting a cell cycle function This gene has been found fused to a variety of translocation partner genes in various leukemias most notably the t(9 22) translocation that results in a fusion with the 5 end of the breakpoint cluster region gene (BCR MIM 151410) Alternative splicing of this gene results in two transcript variants which contain alternative first exons that are spliced to the remaining common exons
Catalog No. 50-000-06829