Learn More
Medchemexpress LLC C-C motif chemokine 17 (TARC), rhesus macaque recombinant protein (C-His) | >95.0% | 50 UG

Supplier: Medchemexpress LLC HYP7666750UG
Recombinant rhesus macaque TARC/CCL17 (A24-S94) is supplied as a C-terminal His-tagged protein expressed in Sf9 insect cells for research on chemokine signaling, T-cell chemotaxis, and CCR4-mediated responses. The product is provided at 50 μg in buffered solution for in vitro research applications.
- Recombinant rhesus macaque sequence A24-S94 (accession Q8HYP9).
- C-terminal His tag for purification and detection.
- Expressed in Sf9 insect cells for proper folding and processing.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Predicted molecular mass approximately 8.9 kDa.
- Supplied in 20 mM Tris, 500 mM NaCl, 10% glycerol, pH 7.5; 0.22 μm filtered.
- Store at -80°C and avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.