Learn More
Invivogen c-di-GMP STING Agonist, 61093-23-0, MFCD31563661, 1mg
SDP
Supplier:  Invivogen TLRLNACDG5
Encompass
Description: STING agonist. Synonym: Cyclic di-guanylate monophosphate, c-di-GMP sodium salt. CAS number: 61093-23-0. Formula: C20H22N10O14P2 .2Na. Molecular weight: 734.38. Purity: ≥ 95% by LC/MS. Solubility: 50 mg/ml in water. Quality control: The biological activity is assessed by measuring induction of the IFN pathway in THP1-Blue™ ISG cells. The absence of endotoxins and other bacterial contaminants has been confirmed using HEK-Blue™ TLR4 and HEK-Blue™ TLR2 cells.
Catalog No. NC1638994