Learn More
Medchemexpress LLC C-erbB-4/HER4 Antibody (YA456) | Affinity purified | 147 kDa | 100 UL
SDP
Supplier:  Medchemexpress LLC HYP80660100UL
Encompass_Preferred
C-erbB-4/HER4 Antibody (YA456) is a Rabbit-derived and non-conjugated IgG monoclonal antibody, targeting C-erbB-4/HER4. This antibody is for research use only and is affinity purified.
  • Rabbit-derived
  • Monoclonal antibody
  • Targets C-erbB-4/HER4
  • Predicted molecular weight of 147 kDa
  • Affinity purified
  • Stores at -20°C for 1 year; avoid repeated freeze/thaw cycles
  • Ships with blue ice
Catalog No. 50-003-95812