Learn More
Sigma Aldrich Fine Chemicals Biosciences C-flexR laboratory tubing
Supplier:  Sigma Aldrich Fine Chemicals Biosciences T791325FT
Encompass
Applications General-purpose lab uses requiring high performance and long life.Characteristics Exceptional tensile and tear strength good biocompatibility excellent chemical resistance low gas permeability and a smooth surface. Opaque white.Certification Resin meets USP Class VI requirements. Tubing is manufactured according to GMP.Temperature range -50 to 275f (-45 to 135C)Sterilize With autoclave ethylene oxide or gamma irradiation.
Catalog No. 50-290-0054