Learn More
Enzo Life Sciences c FLIP polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences ADIAAP440E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of human c-FLIP6. The sequence conserved among all c-FLIP variants. UniProt: O15519. Species Reactivity: Human, Mouse, Rat. Applications: ICC, WB. Recommended dilutions: Immunocytochemistry (5µg/ml). Western Blot (0.5-1.0µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-658-783