Learn More
Medchemexpress LLC C.I. Pigment red 123 | 24108-89-2 | 98.0% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYD0531
Encompass_Preferred
C.I. Pigment red 123 is a significant colorant used to add or alter the color of materials. It is chemically stable and largely unaffected by the carrier or medium in which it is incorporated.
  • Used as a colorant in multiple industries (textile, medicine, food, cosmetics, plastics, paint, ink, photography, paper).
  • Chemically unaffected by the carrier or medium.
Catalog No. 50-004-14301