Learn More
Medchemexpress LLC c-Kit-IN-1 | 1225278-16-9 | MFCD09839958 | C26H21F2N5O3 | 50 MG

Supplier: Medchemexpress LLC HY1524050MG
c-Kit-IN-1 is a potent inhibitor of c-Kit and c-Met with IC50s of less than 200 nM. It also inhibits KDR, PDGFRα, and PDGFRβ. This product is a white to off-white solid with a purity of 98.26% and is designed for research use only.
- Potent inhibitor of c-Kit and c-Met
- Inhibits KDR, PDGFRα, and PDGFRβ
- High purity (98.26%)
- Supplied as a white to off-white solid
- Powder stable at -20°C for 3 years or 4°C for 2 years
- In solvent stable at -80°C for 2 years or -20°C for 1 year
- Soluble in DMSO at 100 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.