Learn More
Sino Biological Recombinant Human c-Kit Protein (D816E, GST Tag), Active
Supplier:  Sino Biological C0612IG500
Encompass_Preferred
Recombinant human c-KIT (D816E) (544-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-5720