Learn More
Genetex Inc C-Peptide Monoclonal Antibody [HMV363] HistoMAX™ (GTX639928) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX639928
Encompass_Preferred
The C-Peptide antibody [HMV363] HistoMAX™ (GTX639928) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human pancreatic neuroendocrine tumor (PNET, insulinoma) tissue using GTX639928 C-Peptide antibody [HMV363] HistoMAX. Strong C-Peptide staining of all tumor cells.
Catalog No. NC3669296