Learn More
Medchemexpress LLC HY-111413A 5mg Medchemexpress, c(phg-isoDGR-(NMe)k) (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY111413A5MG
Encompass_Preferred
Medchemexpress, HY-111413A 5mg c(phg-isoDGR-(NMe)k) (TFA) CAS: c(phg-isoDGR-(NMe)k) TFA is a selective and potent α5β1-integrin ligand with an IC50 of 2.9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8862