Learn More
Carbosynth LLC. Cyclo[Arg-Gly-Asp-D-Phe-Lys(PEG-PEG)], 5mg, MW: (M.W. 894.0), Molecular formula: C39H63N11O13
SDP
Supplier:  Carbosynth LLC. PCI3696PI5MG
Encompass_Preferred
Synonyms: cyclo [Arg-Gly-Asp-D-Phe-Lys(PEG-PEG)], c(RGDfK(PEG-PEG)) where PEG = 8-Amino-3,6-Dioxaoctanoic Acid, Application: RGD Peptide equipped with PEG spacers for more efficient binding to lipid surfaces(Requires further derivatization before use), Storage Temperature: -20°C, References: P. Holig, M. Bach, T. Volkel, T. Nahde, S. Hoffmann, R. Muller, and R.E. Kontermann, Protein Engin. Design Selection, 17, 433 (2004).
Catalog No. 50-168-6290