Learn More
Medchemexpress LLC C004019 | 2417159-57-8 | 99.7% | 1035.29 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY138669100MG
Encompass_Preferred
C004019 is a BBB-penetrable, small-molecule PROTAC designed to target tau protein. It works by recruiting tau and E3 ligase simultaneously, promoting the ubiquitination and proteasome-dependent degradation of tau. This action has been shown to improve synaptic and cognitive functions in Alzheimer's disease (AD) mice, making it suitable for research in AD and tau protein-related diseases.
  • Does not significantly affect viability of HEK293 cells.
  • Inhibits expression of total tau and phosphorylated tau in HEK293-hTau and SH-SY5Y cells.
  • Reduces tau protein levels in the brains of wild-type and 3xTg-AD mice.
  • Promotes tau clearance and improves cognitive and synaptic functions in hTau and 3xTg-AD mice.
  • Ameliorates cognitive functions and dendritic plasticity.
Catalog No. 50-003-52482