Learn More
Cayman Chemical C1 InhIbItr PolycLNal ntIb
SDP
Supplier:  Cayman Chemical 38085200 UL
Encompass_Preferred
An isoflavonoid with antioxidant activity scavenges peroxynitrite nitric oxide radicals and total ROS (IC50s 1.36 1.13 and 6.51 UM respectively)
Catalog No. 50-291-7681