Learn More
Cayman Chemical C10 AV CeramIded18 110 0 1mg
SDP
Supplier:  Cayman Chemical 90007531 MG
Encompass_Preferred
A fluorescently tagged probe consisting of C-10 ceramide with an AV group attached to the end of the acyl chain
Catalog No. 50-289-2650