Learn More
Cayman Chemical C10 CeramIded18 110 0 10mg
SDP
Supplier:  Cayman Chemical 2252910 MG
Encompass_Preferred
A bioactive sphingolipid; inhibits growth of wild-type and MDR1 gene-transfected MDA435/LCC6 human breast cancer cells (IC50s = 44.1 and 46 μM, respectively); inhibits the growth of J774 murine macrophages in a dose-dependent manner
Catalog No. 50-298-6721