Learn More
Cayman Chemical C11 BODIPY 581591 1mg
Supplier: Cayman Chemical 270861 MG
A lipid-soluble ratiometric fluorescent indicator of lipid oxidation; ex/em = 581/591 nm, respectively, when reduced, and 500/510 nm, respectively, when oxidized; has been used in the quantitation of ferroptosis
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.