Learn More
Medchemexpress LLC C12-113 | 1220890-27-6 | 98.6% | 854.47 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1605545MG
Encompass_Preferred
C12-113 is a lipidoid delivery agent that can be used to transfect siRNA into cells. It can also be combined with other lipids to form lipid nanoparticles (LNPs) for the delivery of mRNA encoding the spike glycoprotein of severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) in mice.
  • Lipidoid delivery agent
  • Can transfect siRNA into cells
  • Can be combined with other lipids to form LNPs for mRNA delivery
Catalog No. 50-003-91512