Learn More
Medchemexpress LLC C12-200 | 1220890-25-4 | MFCD32062847 | 99.9% | 1136.93 g/mol | C70H145N5O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1454055MG
Encompass_Preferred
C12-200 is a branched-chain ionizable cationic lipidoid used as a helper lipid in lipid nanoparticle (LNP) formulations for mRNA delivery. It is a high-molecular-weight liquid lipid with documented preclinical use delivering mRNA to produce therapeutic proteins in animal models.
  • Ionizable cationic lipidoid suitable for LNP formulation.
  • Demonstrated mRNA delivery and protein expression in preclinical models.
  • High chemical purity for research applications.
  • Liquid physical state facilitates formulation handling.
  • Well-characterized molecular properties (C70H145N5O5, 1136.93 g/mol).
  • COA and SDS documentation available for quality and safety review.
Catalog No. 50-002-21285