Learn More
TARGETMOL CHEMICALS INC C12-200 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T386045MG
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. C12-200 is a benchmark ionizable cationic lipidoid along with helper lipids and can be used in the delivery of mRNA. Purity 99.51%
Catalog No. 50-287-8638