Learn More
Cayman Chemical C12 CeramIded18 112 0 10mg
SDP
Supplier:  Cayman Chemical 2253010 MG
Encompass_Preferred
A naturally occurring ceramide; is formed when C12 sphingomyelin is hydrolyzed by ASM; substrate for acid ceramidase, an enzyme which has 2- to 12-fold higher activity in fibroblasts from patients with Farber disease; found in the Wharton's jelly of babies born to women with pre-eclampsia
Catalog No. 50-298-6742