Learn More
Medchemexpress LLC C12-iE-DAP hydrochloride | 1269619-57-9 | 538.07 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP5524A5MG
Encompass_Preferred
C12-iE-DAP hydrochloride is the hydrochloride salt form of C12-iE-DAP. This biologically active peptide functions as an agonist for NOD1, formed by adding a lauroyl (C12) group to the glutamic residue of iE-DAP.
  • Biologically active peptide
  • Functions as a NOD1 agonist
  • Targets NOD-like receptor (NLR)
  • Relevant to immunology and inflammation pathways
Catalog No. 50-003-98577