Learn More
Medchemexpress LLC C12-TLRa | 3067466-15-0 | C46H73N5O2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY153879100MG
Encompass_Preferred
C12-TLRa is an ionized adjuvant lipid that targets TLR7/8 receptors in endosomes. It enhances mRNA delivery as a structural component of LNPs and boosts innate immune responses. This substance can be used for the research of SARS-CoV-2 mRNA vaccine.
  • Enhances mRNA delivery as a structural component of LNPs
  • Boosts innate immune responses
  • Targets TLR7/8 receptors in endosomes
  • Can be used for SARS-CoV-2 mRNA vaccine research
Catalog No. 50-003-76779