Learn More
Medchemexpress LLC C12-TLRa | 3067466-15-0 | C46H73N5O2 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY153879
Encompass_Preferred
C12-TLRa is an ionized adjuvant lipid that targets TLR7/8 receptors in endosomes. It enhances mRNA delivery as a structural component of LNPs and enhances innate immune responses. This compound can be used for the research of SARS-CoV-2 mRNA vaccine.
  • Targets TLR7/8 receptors in endosomes
  • Enhances mRNA delivery as a structural component of LNPs
  • Enhances innate immune responses
  • Used for SARS-CoV-2 mRNA vaccine research
Catalog No. 50-003-74573